accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3FFFNH2

CAS RN 771572-43-1

IUPAC Name: 1-[2-methyl-3-(trifluoromethyl)phenyl]methanamine
Synonyms: 1-[2-methyl-3-(trifluoromethyl)phenyl]methanamine
Molecular Weight (g/mol): 189.18
Molecular Formula: C9H10F3N
InChi Key: WXYVXJSOJCMWGN-UHFFFAOYSA-N
SMILES: CC1=C(CN)C=CC=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results