accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 77152-97-7

IUPAC Name: tert-butyl N-(2-carbamothioylethyl)carbamate
Synonyms: tert-butyl N-(2-carbamothioylethyl)carbamate
Molecular Weight (g/mol): 204.29
Molecular Formula: C8H16N2O2S
InChi Key: OBDMXQCRRWGEQM-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCC(N)=S
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results