accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 77123-57-0

CH3SiH3CCH3O

CAS RN 77123-57-0

IUPAC Name: 4-[2-(trimethylsilyl)ethynyl]benzaldehyde
Synonyms: 4-[2-(trimethylsilyl)ethynyl]benzaldehyde
Molecular Weight (g/mol): 202.33
Molecular Formula: C12H14OSi
InChi Key: UZQDUXAJFTWMDT-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CC1=CC=C(C=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results