accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 77119-85-8

CH3H3CCH3OONH(R)O

CAS RN 77119-85-8

IUPAC Name: tert-butyl N-[(2R)-1-oxo-3-phenylpropan-2-yl]carbamate
Synonyms: tert-butyl N-[(2R)-1-oxo-3-phenylpropan-2-yl]carbamate
Molecular Weight (g/mol): 249.31
Molecular Formula: C14H19NO3
InChi Key: ZJTYRNPLVNMVPQ-GFCCVEGCSA-N
SMILES: CC(C)(C)OC(=O)N[C@H](CC1=CC=CC=C1)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)