accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 768-59-2

H3COH

CAS RN 768-59-2

IUPAC Name: (4-ethylphenyl)methanol
Synonyms: 4-ethylbenzyl alcohol
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: YSLBFFIVJGJBSA-UHFFFAOYSA-N
SMILES: CCC1=CC=C(CO)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results