accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 766-83-6

ClHC

CAS RN 766-83-6

IUPAC Name: 1-chloro-3-ethynylbenzene
Synonyms: 1-chloro-3-ethynylbenzene
Molecular Weight (g/mol): 136.58
Molecular Formula: C8H5Cl
InChi Key: GRBJPHPMYOUMJV-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(=C1)C#C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results