accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CN

CAS RN 766-09-6

IUPAC Name: 1-ethylpiperidine
Synonyms: N-ethylpiperidine ephp cpd N-aethylpiperidin
Molecular Weight (g/mol): 113.2
Molecular Formula: C7H15N
InChi Key: HTLZVHNRZJPSMI-UHFFFAOYSA-N
SMILES: CCN1CCCCC1
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)