accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7657-09-2

IUPAC Name: 1,4-dibromo-2-(trifluoromethyl)benzene
Synonyms: 1,4-dibromo-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 303.9
Molecular Formula: C7H3Br2F3
InChi Key: VWKFJAOCLPPQGR-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Br)=CC=C1Br
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results