accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

P

CAS RN 7650-91-1

IUPAC Name: benzyldiphenylphosphane
Synonyms: benzyldiphenylphosphane
Molecular Weight (g/mol): 276.32
Molecular Formula: C19H17P
InChi Key: UZCPNEBHTFYJNY-UHFFFAOYSA-N
SMILES: C(P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)