accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 765-15-1

IUPAC Name: (dodecylsulfanyl)carbonitrile
Synonyms: thiocyanic acid, dodecyl ester
Molecular Weight (g/mol): 227.41
Molecular Formula: C13H25NS
InChi Key: RYZZMYXTNUJGMU-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCSC#N
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)