accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CClO

CAS RN 764-85-2

IUPAC Name: nonanoyl chloride
Synonyms: nonanoyl chloride
Molecular Weight (g/mol): 176.68
Molecular Formula: C9H17ClO
InChi Key: NTQYXUJLILNTFH-UHFFFAOYSA-N
SMILES: CCCCCCCCC(Cl)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results