accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3H2NH2NBr

CAS RN 76153-06-5

IUPAC Name: 5-bromo-3-methylbenzene-1,2-diamine
Synonyms: 5-bromo-3-methylbenzene-1,2-diamine
Molecular Weight (g/mol): 201.07
Molecular Formula: C7H9BrN2
InChi Key: UOFSLKHZOPVGHG-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CC(N)=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results