accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 76006-33-2

CH3BrOOH

CAS RN 76006-33-2

IUPAC Name: 3-bromo-2-methylbenzoic acid
Synonyms: 3-bromo-2-methylbenzoic acid
Molecular Weight (g/mol): 215.05
Molecular Formula: C8H7BrO2
InChi Key: BJGKVCKGUBYULR-UHFFFAOYSA-N
SMILES: CC1=C(C=CC=C1Br)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)