CAS RN 760-20-3
CAS RN 760-20-3
IUPAC Name:
3-methylpent-1-ene
Synonyms:
3-methyl-1-pentene
Molecular Weight (g/mol):
84.16
Molecular Formula:
C6H12
InChi Key:
LDTAOIUHUHHCMU-UHFFFAOYNA-N
SMILES:
CCC(C)C=C
1
–
1
of
1
results
1
3-Methyl-1-pentene, 98%, Thermo Scientific Chemicals
CAS: 760-20-3 Molecular Formula: C6H12 Molecular Weight (g/mol): 84.162 MDL Number: MFCD00009339 InChI Key: LDTAOIUHUHHCMU-UHFFFAOYSA-N PubChem CID: 12969 IUPAC Name: 3-methylpent-1-ene SMILES: CCC(C)C=C