CAS RN 76-87-9
CAS RN 76-87-9
IUPAC Name:
triphenylstannanol
Synonyms:
triphenyltin hydroxide
Molecular Weight (g/mol):
369.05
Molecular Formula:
C18H18OSn
InChi Key:
ZJIGGMIMCKZRRB-UHFFFAOYSA-N
SMILES:
O.C1=CC=C(C=C1)[SnH](C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Triphenyltin hydroxide, Thermo Scientific™
CAS: 76-87-9 Molecular Formula: C18H18OSn Molecular Weight (g/mol): 369.05 MDL Number: MFCD00013928 InChI Key: ZJIGGMIMCKZRRB-UHFFFAOYSA-N Synonym: triphenyltin hydroxide,fentin hydroxide,triphenylstannanol,vancide ks,hydroxytriphenyltin,hydroxytriphenylstannane,erithane,fenolovo,tenhide,stannane, hydroxytriphenyl PubChem CID: 6327657 IUPAC Name: triphenyltin;hydrate SMILES: O.C1=CC=C(C=C1)[SnH](C1=CC=CC=C1)C1=CC=CC=C1