accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

LiBH3COOCH3H3COOCH3H3COOCH3

CAS RN 75965-35-4

IUPAC Name: lithium(1+) tris(1,2-dimethoxyethane) tetraphenylboranuide
Synonyms: lithium(1+) tris(1,2-dimethoxyethane) tetraphenylborate
Molecular Weight (g/mol): 596.54
Molecular Formula: C36H50BLiO6
InChi Key: ADYUZFWVPWDPFK-UHFFFAOYSA-N
SMILES: [Li+].COCCOC.COCCOC.COCCOC.C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results