CAS RN 758-12-3
IUPAC Name:
acetate
Synonyms:
acetate caswell no. 003 glacial acetate vinegar (substance) azetat acetic acid product (product) fema number 2006 ethanoat acetic acid,glacial ion(1-) acetic acid
Molecular Weight (g/mol):
59.05
Molecular Formula:
C2H3O2
InChi Key:
QTBSBXVTEAMEQO-UHFFFAOYSA-M
SMILES:
CC([O-])=O
1
–
4
of
4
results
2
Acetic acid-d, for NMR, 98 atom % D
CAS: 758-12-3 Molecular Formula: C2H4O2 Molecular Weight (g/mol): 61.058 MDL Number: MFCD00051050 InChI Key: QTBSBXVTEAMEQO-DYCDLGHISA-N Synonym: acetic acid-d,acetic acid-d1,monodeuteroacetic acid,acetic 2h acid,acetic acid-od,o-2h acetic acid,mono-deuteroacetic acid,ch3cood PubChem CID: 2723902 IUPAC Name: deuterio acetate SMILES: CC(=O)O
3
Acetic Acid-d, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Acetic Acid-d, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.