accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 75747-14-7

H3CO(R)(R)H3CNHH2CONHO(R)OCH3(R)ONH2O(R)CH3(S)OHCH3H3CO(Z)(E)(E)

CAS RN 75747-14-7

IUPAC Name: (4Z,6E,8R,9R,10E,12R,13S,14R,16R)-13-hydroxy-8,14-dimethoxy-4,10,12,16-tetramethyl-3,20,22-trioxo-19-[(prop-2-en-1-yl)amino]-2-azabicyclo[16.3.1]docosa-1(21),4,6,10,18-pentaen-9-yl carbamate
Synonyms: tanespimycin
Molecular Weight (g/mol): 585.7
Molecular Formula: C31H43N3O8
InChi Key: AYUNIORJHRXIBJ-CQFHKJEWSA-N
SMILES: CO[C@@H]1C[C@H](C)CC2=C(NCC=C)C(=O)C=C(NC(=O)\C(C)=C/C=C/[C@@H](OC)[C@H](OC(N)=O)\C(C)=C\[C@@H](C)[C@@H]1O)C2=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (4)
  • (3)
Physical Form 
  • (4)
  • (3)
Color 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results