accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 75629-57-1

NClO

CAS RN 75629-57-1

IUPAC Name: 1-[(4-chlorophenyl)methyl]-1H-indole-3-carbaldehyde
Synonyms: 1-[(4-chlorophenyl)methyl]indole-3-carbaldehyde
Molecular Weight (g/mol): 269.73
Molecular Formula: C16H12ClNO
InChi Key: ZDRQMXCSSAPUMM-UHFFFAOYSA-N
SMILES: ClC1=CC=C(CN2C=C(C=O)C3=CC=CC=C23)C=C1
Quantity 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)