accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3CH3H3CCl

CAS RN 753-89-9

IUPAC Name: 1-chloro-2,2-dimethylpropane
Synonyms: 1-chloro-2,2-dimethylpropane
Molecular Weight (g/mol): 106.59
Molecular Formula: C5H11Cl
InChi Key: JEKYMVBQWWZVHO-UHFFFAOYSA-N
SMILES: CC(C)(C)CCl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)