accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 75224-64-5

ONSN

CAS RN 75224-64-5

IUPAC Name: 6-phenyl-2H,3H-imidazo[2,1-b][1,3]thiazole-5-carbaldehyde
Synonyms: 6-phenyl-2H,3H-imidazo[2,1-b][1,3]thiazole-5-carbaldehyde
Molecular Weight (g/mol): 230.29
Molecular Formula: C12H10N2OS
InChi Key: XOGXXJXDZCAAPL-UHFFFAOYSA-N
SMILES: O=CC1=C(N=C2SCCN12)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results