accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H2NNHOONOO

CAS RN 75129-74-7

IUPAC Name: 2-(4-nitrophenoxy)acetohydrazide
Synonyms: 2-(4-nitrophenoxy)acetohydrazide
Molecular Weight (g/mol): 211.18
Molecular Formula: C8H9N3O4
InChi Key: BDPKZWIKLVUGKS-UHFFFAOYSA-N
SMILES: NNC(=O)COC1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results