accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OFFF

CAS RN 75091-99-5

IUPAC Name: 4-(trifluoromethyl)cyclohexan-1-one
Synonyms: 4-(trifluoromethyl)cyclohexan-1-one
Molecular Weight (g/mol): 166.14
Molecular Formula: C7H9F3O
InChi Key: IPQDZFUZVJKAKZ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1CCC(=O)CC1
Quantity 
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (1)
  • (2)