accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 75-04-7

H3CNH2

CAS RN 75-04-7

IUPAC Name: ethanamine
Synonyms: ethylamine 14C-labeled ethylamine ethylamine, 14C-labeled aethylamine ethylamine, 14c-labeled etyloamina etilamina ethylamine (substance) 14c-labeled ethylamine
Molecular Weight (g/mol): 45.09
Molecular Formula: C2H7N
InChi Key: QUSNBJAOOMFDIB-UHFFFAOYSA-N
SMILES: CCN
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)