accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HNSOOClOHO

CAS RN 749884-42-2

IUPAC Name: 3-(3-chlorobenzenesulfonamido)benzoic acid
Synonyms: 3-(3-chlorobenzenesulfonamido)benzoic acid
Molecular Weight (g/mol): 311.74
Molecular Formula: C13H10ClNO4S
InChi Key: APBOVLPLJFJSRI-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(NS(=O)(=O)C2=CC(Cl)=CC=C2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results