accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 749875-16-9

IUPAC Name: 5-bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid
Synonyms: 5-bromo-2-(trifluoromethyl)pyridine-4-carboxylic acid
Molecular Weight (g/mol): 270.01
Molecular Formula: C7H3BrF3NO2
InChi Key: XNVAVQMVAYUTBA-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=NC=C1Br)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results