accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NBrF

CAS RN 749269-73-6

IUPAC Name: 1-(4-bromo-3-fluorophenyl)cyclopropane-1-carbonitrile
Synonyms: 1-(4-bromo-3-fluorophenyl)cyclopropane-1-carbonitrile
Molecular Weight (g/mol): 240.08
Molecular Formula: C10H7BrFN
InChi Key: KVWJVBSLLYGPIM-UHFFFAOYSA-N
SMILES: FC1=C(Br)C=CC(=C1)C1(CC1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results