accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7473-93-0

IUPAC Name: (2E)-3-(2-nitrophenyl)-1-phenylprop-2-en-1-one
Synonyms: (2E)-3-(2-nitrophenyl)-1-phenylprop-2-en-1-one
Molecular Weight (g/mol): 253.26
Molecular Formula: C15H11NO3
InChi Key: KTXHLWZQKQDFRF-ZHACJKMWSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1\C=C\C(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results