accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7465-87-4

H3COOHN

CAS RN 7465-87-4

IUPAC Name: N-benzyl-4-methoxybenzamide
Synonyms: N-benzyl-4-methoxybenzamide
Molecular Weight (g/mol): 241.29
Molecular Formula: C15H15NO2
InChi Key: GEFZSLMGZREDTO-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)NCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results