CAS RN 74507-63-4
CAS RN 74507-63-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
329.16
Molecular Formula:
C20H30Ni
InChi Key:
JLYFAOFYDLSGOE-UHFFFAOYSA-N
SMILES:
Cc1c(C)c(C)[c-](C)c1C.C[C-]12[C-]3(C)[C-]4(C)[C-]5(C)[C-]1(C)[Ni]2345
1
–
1
of
1
results
1
Bis(pentamethylcyclopentadienyl)nickel
CAS: 74507-63-4 Molecular Formula: C20H30Ni Molecular Weight (g/mol): 329.153 MDL Number: MFCD00058704 InChI Key: CHPLEWYRKUFKQP-UHFFFAOYSA-N Synonym: Decamethylnickelocene PubChem CID: 24943158 IUPAC Name: nickel(2+);1,2,3,5,5-pentamethylcyclopenta-1,3-diene SMILES: CC1=[C-]C(C(=C1C)C)(C)C.CC1=[C-]C(C(=C1C)C)(C)C.[Ni+2]