accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NCH3OSiCH3CH3H3C

CAS RN 7449-74-3

IUPAC Name: N-methyl-N-(trimethylsilyl)acetamide
Synonyms: N-methyl-N-(trimethylsilyl)acetamide
Molecular Weight (g/mol): 145.28
Molecular Formula: C6H15NOSi
InChi Key: QHUOBLDKFGCVCG-UHFFFAOYSA-N
SMILES: CN(C(C)=O)[Si](C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)