accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7446-31-3

H2OH2OH2OH2OOSOOOOSOOOZr

CAS RN 7446-31-3

IUPAC Name: zirconium(4+) tetrahydrate disulfate
Synonyms: zirconium(4+) tetrahydrate disulfate
Molecular Weight (g/mol): 355.4
Molecular Formula: H8O12S2Zr
InChi Key: NLOQZPHAWQDLQW-UHFFFAOYSA-J
SMILES: O.O.O.O.[Zr+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)