CAS RN 7423-96-3
CAS RN 7423-96-3
IUPAC Name:
2-amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid
Synonyms:
pardinon fluorthyrin m-ftyr
Molecular Weight (g/mol):
199.18
Molecular Formula:
C9H10FNO3
InChi Key:
VIIAUOZUUGXERI-UHFFFAOYNA-N
SMILES:
NC(CC1=CC(F)=C(O)C=C1)C(O)=O
1
–
1
of
1
results
1
3-Fluoro-L-tyrosine, 97%, Thermo Scientific Chemicals
CAS: 7423-96-3 Molecular Formula: C9H10FNO3 Molecular Weight (g/mol): 199.18 MDL Number: MFCD00002607 InChI Key: VIIAUOZUUGXERI-JLDDOWRYNA-N Synonym: 3-fluoro-l-tyrosine,s-2-amino-3-3-fluoro-4-hydroxyphenyl propanoic acid,3-fluorotyrosine,3-fluorotyrosine, l,h-tyr 3-f-oh,unii-174nrg3m2x,s-3-fluorotyrosine,3'-fluoro-l-tyrosine,2s-2-amino-3-3-fluoro-4-hydroxyphenyl propanoic acid,3-fluoro-tyrosine PubChem CID: 643330 ChEBI: CHEBI:46534 IUPAC Name: (2S)-2-amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid SMILES: N[C@@H](CC1=CC=C(O)C(F)=C1)C(O)=O