accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ONOOHO

CAS RN 7423-55-4

IUPAC Name: 3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanoic acid
Synonyms: 3-(2,5-dioxopyrrol-1-yl)propanoic acid
Molecular Weight (g/mol): 169.14
Molecular Formula: C7H7NO4
InChi Key: IUTPJBLLJJNPAJ-UHFFFAOYSA-N
SMILES: OC(=O)CCN1C(=O)C=CC1=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)