accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7412-78-4

H2NONHOHOOHO

CAS RN 7412-78-4

IUPAC Name: 2-(2-aminoacetamido)pentanedioic acid
Synonyms: 2-(2-aminoacetamido)pentanedioic acid
Molecular Weight (g/mol): 204.18
Molecular Formula: C7H12N2O5
InChi Key: IEFJWDNGDZAYNZ-UHFFFAOYNA-N
SMILES: NCC(=O)NC(CCC(O)=O)C(O)=O
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results