accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CH3CCH3OBOCH3H3CCH3OCH3H3CCH3

CAS RN 7397-43-5

IUPAC Name: tri-tert-butyl borate
Synonyms: tri-tert-butyl borate
Molecular Weight (g/mol): 230.16
Molecular Formula: C12H27BO3
InChi Key: ZMCWFMOZBTXGKI-UHFFFAOYSA-N
SMILES: CC(C)(C)OB(OC(C)(C)C)OC(C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)