accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7364-25-2

IUPAC Name: 2,3-dihydro-1H-indazol-3-one
Synonyms: 1,2-dihydroindazol-3-one
Molecular Weight (g/mol): 134.14
Molecular Formula: C7H6N2O
InChi Key: SWEICGMKXPNXNU-UHFFFAOYSA-N
SMILES: O=C1NNC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results