accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CONClO

CAS RN 73568-29-3

IUPAC Name: 2-chloro-6-methoxyquinoline-3-carbaldehyde
Synonyms: 2-chloro-6-methoxyquinoline-3-carbaldehyde
Molecular Weight (g/mol): 221.64
Molecular Formula: C11H8ClNO2
InChi Key: TZQOMBXDCIPJKW-UHFFFAOYSA-N
SMILES: COC1=CC=C2N=C(Cl)C(C=O)=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results