accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OOOOH3CNOO

CAS RN 73357-18-3

IUPAC Name: 2-(4,5-dimethoxy-2-nitrophenyl)acetate
Synonyms: 2-(4,5-dimethoxy-2-nitrophenyl)acetate
Molecular Weight (g/mol): 240.19
Molecular Formula: C10H10NO6
InChi Key: WJPMJFUEMVXUCV-UHFFFAOYSA-M
SMILES: COC1=CC(CC([O-])=O)=C(C=C1OC)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results