accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7335-27-5

H3COOCl

CAS RN 7335-27-5

IUPAC Name: ethyl 4-chlorobenzoate
Synonyms: ethyl 4-chlorobenzoate
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: RWBYCMPOFNRISR-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(Cl)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results