accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 7328-91-8

CH3H3CNH2H2N

CAS RN 7328-91-8

IUPAC Name: 2,2-dimethylpropane-1,3-diamine
Synonyms: 2,2-dimethylpropane-1,3-diamine
Molecular Weight (g/mol): 102.18
Molecular Formula: C5H14N2
InChi Key: DDHUNHGZUHZNKB-UHFFFAOYSA-N
SMILES: CC(C)(CN)CN
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results