accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OOOO

CAS RN 7325-46-4

IUPAC Name: 2-[4-(carboxylatomethyl)phenyl]acetate
Synonyms: 2-[4-(carboxylatomethyl)phenyl]acetate
Molecular Weight (g/mol): 192.17
Molecular Formula: C10H8O4
InChi Key: SLWIPPZWFZGHEU-UHFFFAOYSA-L
SMILES: [O-]C(=O)CC1=CC=C(CC([O-])=O)C=C1
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results