accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOCH3CH3H3C

CAS RN 7307-04-2

IUPAC Name: 5,5-dimethylhexane-2,4-dione
Synonyms: 5,5-dimethyl-2,4-hexanedione
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: LCLCVVVHIPPHCG-UHFFFAOYSA-N
SMILES: CC(=O)CC(=O)C(C)(C)C
Boiling Point 
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results