accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 73035-16-2

H3COSOOCl

CAS RN 73035-16-2

IUPAC Name: 3-acetylbenzene-1-sulfonyl chloride
Synonyms: 3-acetylbenzenesulfonyl chloride
Molecular Weight (g/mol): 218.65
Molecular Formula: C8H7ClO3S
InChi Key: CGMBNEIGZOCPPP-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(=CC=C1)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results