accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClSOONHOOH

CAS RN 727689-55-6

IUPAC Name: 4-(3-chlorobenzenesulfonamido)benzoic acid
Synonyms: 4-(3-chlorobenzenesulfonamido)benzoic acid
Molecular Weight (g/mol): 311.74
Molecular Formula: C13H10ClNO4S
InChi Key: JQVIQJNDKGSTPV-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(NS(=O)(=O)C2=CC(Cl)=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results