accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 72768-95-7

OFFBr

CAS RN 72768-95-7

IUPAC Name: 1-(bromomethyl)-3-(difluoromethoxy)benzene
Synonyms: 1-(bromomethyl)-3-(difluoromethoxy)benzene
Molecular Weight (g/mol): 237.04
Molecular Formula: C8H7BrF2O
InChi Key: TZYKSTKEVLDYAP-UHFFFAOYSA-N
SMILES: FC(F)OC1=CC=CC(CBr)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results