accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 725702-35-2

CH3OSH2N

CAS RN 725702-35-2

IUPAC Name: 2-ethoxybenzene-1-carbothioamide
Synonyms: 2-ethoxybenzenecarbothioamide
Molecular Weight (g/mol): 181.25
Molecular Formula: C9H11NOS
InChi Key: FUQRHZQJAIHJQB-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1C(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results