accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HOOHNBr

CAS RN 7254-19-5

IUPAC Name: 5-bromo-1H-indole-2-carboxylic acid
Synonyms: 5-bromo-1H-indole-2-carboxylic acid
Molecular Weight (g/mol): 240.06
Molecular Formula: C9H6BrNO2
InChi Key: YAULOOYNCJDPPU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=CC(Br)=CC=C2N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results