accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHOFFFFFF

CAS RN 725-89-3

IUPAC Name: 3,5-bis(trifluoromethyl)benzoic acid
Synonyms: 3,5-bis(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 258.12
Molecular Formula: C9H4F6O2
InChi Key: HVFQJWGYVXKLTE-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)