accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 72411-52-0

IUPAC Name: 5-(4-fluorophenyl)-1H-pyrazol-3-amine
Synonyms: 5-(4-fluorophenyl)-1H-pyrazol-3-amine
Molecular Weight (g/mol): 177.18
Molecular Formula: C9H8FN3
InChi Key: QYEHDCXFXONDPV-UHFFFAOYSA-N
SMILES: NC1=NNC(=C1)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results